Sodium sulfite, anhydrous ≥98%
Supplier: BeanTown Chemical
145385-10KG
145385-2.5KG
145385-500G
BT145385-10KGEA
288
USD
BT145385-10KG
BT145385-2.5KG
BT145385-500G
Sodium sulfite, anhydrous ≥98%
Sodium sulphite
CAS: 7757-83-7; EC No: 231-821-4; MDL No: MFCD00003503; RTECS: WE2150000
Solid; Linear Formula: Na2SO3; MW: 126.04
Melting Point:
- RTECS: WE2150000
- Air Sensitive, Moisture Sensitive
- Solid
Formula:
Na₂SO₃ Melting Pt: >500 °C (dec.) Density: 2.63 g/mL |
MDL Number:
MFCD00003503 CAS Number: 7757-83-7 EINECS: 231-821-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...