Magnesium sulfate heptahydrate ≥99%
Supplier: BeanTown Chemical
145380-1KG
145380-5KG
BT145380-1KGEA
46.6
USD
BT145380-1KG
BT145380-5KG
Magnesium sulfate heptahydrate ≥99%
Magnesium sulphate heptahydrate
CAS: 10034-99-8; EC No: 231-298-2; MDL No: MFCD00149785; RTECS: OM4508000
Crystalline/Powder; Linear Formula: MgSO4·7H2O; MW: 246.48
Density (g/mL): 1.67
- RTECS: OM4508000
- Crystalline/Powder
Formula:
MgSO₄·7H₂O Density: 1.67 g/mL |
MDL Number:
MFCD00149785 CAS Number: 10034-99-8 EINECS: 231-298-2 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...