Terbium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.62 mm

Supplier: BeanTown Chemical

144895-50X50MM 144895-25X25MM
BT144895-25X25EA 313.44 USD
BT144895-25X25 BT144895-50X50
Terbium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.62 mm
Terbium

  • Solid

Formula: Tb
MW: 158.93 g/mol
Boiling Pt: 3230 °C
Melting Pt: 1356 °C
Density: 8.234 g/mL
MDL Number: MFCD00011256
CAS Number: 7440-27-9
EINECS: 231-137-6

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR