2-Amino-5-iodopyridine ≥97%
Supplier: BeanTown Chemical
Synonyms:
5-Iodopyridin-2-amine, 5-Iodo-2-pyridylamine
144845-1G
144845-25G
144845-5G
BT144845-1GEA
44
USD
BT144845-1G
BT144845-25G
BT144845-5G
2-Amino-5-iodopyridine ≥97%
2-Amino-5-iodopyridine
CAS: 20511-12-0; MDL No: MFCD00160312
Solid; Molecular Formula: C5H5IN2 ; MW: 220.01
Melting Point: 128-131°
Light Sensitive
- Light Sensitive
- Solid
Formula:
C₅H₅IN₂ Melting Pt: 128…131 °C |
MDL Number:
MFCD00160312 CAS Number: 20511-12-0 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...