Sulfolane ≥99%

Supplier: BeanTown Chemical

Synonyms: Tetramethylene sulfone, Tetrahydrothiophene-1,1-dioxide, Tetramethylene sulphone, Tetrahydrothiophene-1,1-dioxide, Tetrahydrothiophene 1,1-dioxide

144390-1KG 144390-250G 144390-5KG
BT144390-1KGEA 124.42 USD
BT144390-1KG BT144390-250G BT144390-5KG
Sulfolane ≥99%
Sulpholane

CAS: 126-33-0; EC No: 204-783-1; MDL No: MFCD00005484; RTECS: XN0700000 Low Melting Solid; Molecular Formula: C4H8O2S; MW: 120.17 Melting Point: 20-26°; Boiling Point: 285°; Flash point: 166°C (330°F) Density (g/mL): 1.261; Refractive Index: 1.484 Hygroscopic


  • RTECS: XN0700000
  • Refractive index: 1.484
  • Hygroscopic
  • Low Melting Solid

Formula: C₄H₈O₂S
Boiling Pt: 285 °C
Melting Pt: 20…26 °C
Density: 1.261 g/mL
Flash Pt: 166 °C (330 °F)
MDL Number: MFCD00005484
CAS Number: 126-33-0
EINECS: 204-783-1

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR