Disodium hydrogen phosphate heptahydrate 98.0-102.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Disodium phosphate (heptahydrate), Disodium hydrogenorthophosphate 7-hydrate, Sodium phosphate dibasic 7-hydrate, Sodium monohydrogen phosphate 7-hydrate, Sodium phosphate dibasic heptahydrate, Disodium hydrogenorthophosphate heptahydrate, di-Sodium hydrogen phosphate 7-hydrate, Sodium hydrogen phosphate heptahydrate, Sodium monohydrogen phosphate heptahydrate
144355-2KG
144355-500G
BT144355-2KGEA
161
USD
BT144355-2KG
BT144355-500G
Disodium hydrogen phosphate heptahydrate 98.0-102.0% ACS
di-Sodium hydrogen phosphate heptahydrate
CAS: 7782-85-6; EC No: 231-448-7; MDL No: MFCD00149180; RTECS: WC4600000
Crystalline/Powder; Linear Formula: Na2HPO4·7H2O; MW: 268.07
Density (g/mL): 1.68
- RTECS: WC4600000
- Crystalline/Powder
Formula:
HNa₂O₄P·7H₂O Density: 1.68 g/mL |
MDL Number:
MFCD00149180 CAS Number: 7782-85-6 EINECS: 231-448-7 |
Specification Test Results
Assay | 98.0-102.0% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...