Melamine ≥99%

Supplier: BeanTown Chemical

144205-10KG 144205-2.5KG 144205-500G
BT144205-10KGEA 187 USD
BT144205-10KG BT144205-2.5KG BT144205-500G
Melamine ≥99%
Melamine

CAS: 108-78-1; EC No: 203-615-4; MDL No: MFCD00006055; RTECS: OS0700000 Solid; Molecular Formula: C3H6N6; MW: 126.12 Melting Point: 300°; Flash point: 300°C (572°F) Density (g/mL): 1.573


  • RTECS: OS0700000
  • Solid

Formula: C₃H₆N₆
Melting Pt: >300 °C
Density: 1.573 g/mL
Flash Pt: 300 °C (572 °F)
MDL Number: MFCD00006055
CAS Number: 108-78-1
EINECS: 203-615-4

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR