1-Chloro-3-methyl-5-nitrobenzene
Supplier: BeanTown Chemical
Synonyms:
3-Chloro-5-nitrotoluene
143985-1G
143985-5G
BT143985-1GEA
49
USD
BT143985-1G
BT143985-5G
1-Chloro-3-methyl-5-nitrobenzene
1-Chloro-3-methyl-5-nitrobenzene
CAS: 16582-38-0
Solid; Linear Formula: ClC6H3(CH3)(NO2); Molecular Formula: C7H6ClNO2; MW: 171.58
Melting Point: 54-55°; Boiling Point: 254°; Flash point: 107°C (225°F)
Density (g/mL): 1.324
- Solid
Formula:
C₇H₆ClNO₂ Boiling Pt: 254 °C Melting Pt: 54…55 °C Density: 1.324 g/mL Flash Pt: 107 °C (225 °F) |
MDL Number:
MFCD09955417 CAS Number: 16582-38-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...