2-Aminopyridine-4-methanol ≥97%
Supplier: BeanTown Chemical
Synonyms:
2-Amino-4-pyridinylmethanol, (2-Aminopyridin-4-yl)-methanol, (2-Aminopyridin-4-yl)methanol, (2-Aminopyrid-4-yl)methanol
143960-1G
143960-5G
BT143960-1GEA
88
USD
BT143960-1G
BT143960-5G
2-Aminopyridine-4-methanol ≥97%
2-Aminopyridine-4-methanol
CAS: 105250-17-7; MDL No: MFCD03791261
Crystalline; Molecular Formula: C6H8N2O; MW: 124.14
Melting Point: 80-84°
- Crystalline
Formula:
C₆H₈N₂O MW: 124.143 g/mol Melting Pt: 80…84 °C |
MDL Number:
MFCD03791261 CAS Number: 105250-17-7 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...