Ethyl benzoate ≥99%

Supplier: BeanTown Chemical

Synonyms: Benzoic acid ethyl ester

143935-100G 143935-2.5KG 143935-500G
BT143935-100GEA 28.3 USD
BT143935-100G BT143935-2.5KG BT143935-500G
Ethyl benzoate ≥99%
Ethyl benzoate

CAS: 93-89-0; EC No: 202-284-3; MDL No: MFCD00009109; RTECS: DH0200000 Liquid; Linear Formula: C6H5COOCH2CH3; Molecular Formula: C9H10O2; MW: 150.18 Melting Point: -34°; Boiling Point: 212°; Flash point: 93°C (199°F) Density (g/mL): 1.045; Refractive Index: 1.504


  • RTECS: DH0200000
  • Refractive index: 1.504
  • Liquid

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 415
Melting Pt: -35
Density: 1.045 g/mL
Flash Pt: 93 °C (199 °F)
MDL Number: MFCD00009109
CAS Number: 93-89-0
EINECS: 202-284-3
Merck Index: 13,03802

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR