8-Bromoisoquinoline

Supplier: BeanTown Chemical

143925-1G 143925-5G
BT143925-1GEA 67 USD
BT143925-1G BT143925-5G
8-Bromoisoquinoline
8-Bromoisoquinoline

CAS: 63927-22-0; MDL No: MFCD04973298 Solid; Molecular Formula: C9H6BrN; MW: 208.05


  • Solid

Formula: C₉H₆BrN
MDL Number: MFCD04973298
CAS Number: 63927-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR