Potassium thiocyanate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Potassium rhodanide
143810-2.5KG
143810-500G
BT143810-2.5KGEA
217.18
USD
BT143810-2.5KG
BT143810-500G
Potassium thiocyanate ≥98%
Potassium thiocyanate
CAS: 333-20-0; EC No: 206-370-1; MDL No: MFCD00011413; RTECS: XL1925000
Crystalline; Linear Formula: KSCN; MW: 97.18
Melting Point: 173°; Boiling Point: 500° (decomposes)
Density (g/mL): 1.89
Light Sensitive, Moisture Sensitive
- RTECS: XL1925000
- Light Sensitive, Moisture Sensitive
- Crystalline
Formula:
KSCN MW: 97.18 g/mol Boiling Pt: 500 (dec.) Melting Pt: 175 Density: 1.89 g/mL |
MDL Number:
MFCD00011413 CAS Number: 333-20-0 EINECS: 206-370-1 Merck Index: 12,07866 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...