Sodium thiosulfate pentahydrate 99

Supplier: BeanTown Chemical

143715-10KG 143715-2.5KG 143715-500G
BT143715-10KGEA 212 USD
BT143715-10KG BT143715-2.5KG BT143715-500G
Sodium thiosulfate pentahydrate 99
Sodium thiosulphate pentahydrate

CAS: 10102-17-7; EC No: 231-867-5; MDL No: MFCD00149186; RTECS: WE6660000 Crystalline; Linear Formula: Na2S2O3·5H2O; MW: 248.17 Melting Point: 48° Density (g/mL): 1.69


  • RTECS: WE6660000
  • Crystalline

Formula: Na₂S₂O₃.5H₂O
MW: 248.19 g/mol
Melting Pt: 48
Density: 1.69 g/mL
MDL Number: MFCD00149186
CAS Number: 10102-17-7
EINECS: 231-867-5
Merck Index: 12,08844

Order Now

Specification Test Results

Assay 99% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR