Barium sulfate ≥97%
Supplier: BeanTown Chemical
143640-5KG
143640-1KG
BT143640-5KGEA
244.3
USD
BT143640-5KG
BT143640-1KG
Barium sulfate ≥97%
Barium sulphate
- 1-4 Micron Powder
Formula:
BaSO₄ MW: 233.39 g/mol |
MDL Number:
MFCD00003455 CAS Number: 7727-43-7 EINECS: 231-784-4 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...