Urea 99.0-100.5% ACS

Supplier: BeanTown Chemical

143530-100G 143530-2KG 143530-500G
BT143530-100GEA 33.28 USD
BT143530-100G BT143530-2KG BT143530-500G
Urea 99.0-100.5% ACS
Urea

CAS: 57-13-6; EC No: 200-315-5; MDL No: MFCD00008022; RTECS: YR6250000 Crystalline; Linear Formula: NH2CONH2; MW: 60.06 Melting Point: 132-135° Density (g/mL): 1.335


  • RTECS: YR6250000
  • Crystalline

Formula: NH₂CONH₂
Melting Pt: 132…135 °C
Density: 1.335 g/mL
MDL Number: MFCD00008022
CAS Number: 57-13-6
EINECS: 200-315-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0-100.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR