Silver ≥99.9%, wool

Supplier: BeanTown Chemical

143510-1G 143510-25G 143510-5G
BT143510-1GEA 41 USD
BT143510-1G BT143510-25G BT143510-5G
Silver ≥99.9%, wool
Silver

CAS: 7440-22-4; EC No: 231-131-3; MDL No: MFCD00003397; RTECS: VW3500000 Solid, approx. 0.05mm dia fibers; Molecular Formula: Ag; MW: 107.87 Melting Point: 960°; Boiling Point: 2212° Density (g/mL): 10.49


  • RTECS: VW3500000
  • Solid, approx. 0.05mm dia fibers

Formula: Ag
Boiling Pt: 2212 °C
Melting Pt: 960 °C
Density: 10.49 g/mL
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR