Guanidinium thiocyanate ≥99%

Supplier: BeanTown Chemical

Synonyms: Guanidinium sulfocyanide, Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide

143285-100G 143285-2.5KG 143285-500G
BT143285-100GEA 69.33 USD
BT143285-100G BT143285-2.5KG BT143285-500G
Guanidinium thiocyanate ≥99%
Guanidinium thiocyanate

CAS: 593-84-0; EC No: 209-812-1; MDL No: MFCD00013027 Crystalline/Powder; Molecular Formula: CH5N3·HSCN; MW: 118.16 Melting Point: 120-122°


  • Crystalline/Powder

Formula: CH₅N₃·CHNS
Melting Pt: 120…122 °C
MDL Number: MFCD00013027
CAS Number: 593-84-0
EINECS: 209-812-1

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR