Platinum ≥99.95% (trace metals basis), wire Ø 0.6 mm, hard type
Supplier: BeanTown Chemical
Synonyms:
Platinium
143250-10CM
143250-50CM
BT143250-10CMEA
256.74
USD
BT143250-10CM
BT143250-50CM
Platinum ≥99.95% (trace metals basis), wire Ø 0.6 mm, hard type
Platinum
CAS: 7440-06-4; EC No: 231-116-1; MDL No: MFCD00011179; RTECS: TP2160000
Solid; Molecular Formula: Pt; MW: 195.08
Melting Point: 1772°; Boiling Point: 3827°
Density (g/mL): 21.45
- RTECS: TP2160000
- Solid
Formula:
Pt Boiling Pt: 3827 °C Melting Pt: 1772 °C Density: 21.45 g/mL |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 EINECS: 231-116-1 |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...