Calcium carbonate ≥99.0% ACS low in alkali

Supplier: BeanTown Chemical

Synonyms: Marble

143200-1KG 143200-250G
BT143200-1KGEA 139 USD
BT143200-1KG BT143200-250G
Calcium carbonate ≥99.0% ACS low in alkali
Calcium carbonate

CAS: 471-34-1; EC No: 207-439-9; MDL No: MFCD00010906; RTECS: FF9335000 Powder; Linear Formula: CaCO3; MW: 100.09 Density (g/mL): 2.93


  • RTECS: FF9335000
  • Powder

Formula: CaCO₃
Density: 2.93 g/mL
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9

Order Now

Specification Test Results

Assay 99.0% min.
Insoluble in dilute HCl ≤0.01%
Ammonium hydroxide PPT ≤0.01%
Chloride (Cl) ≤0.001%
Sulfate (SO4) ≤0.005%
Aluminum (Al) ≤0.001%
Ammonium (NH4) ≤0.003%
Barium (Ba) ≤0.01%
Fluoride (F) ≤0.0005%
Lead (Pb) ≤0.001%
Iron (Fe) ≤0.0007%
Magnesium (Mg) ≤0.01%
Potassium (K) ≤0.01%
Sodium (Na) ≤0.01%
Strontium (Sr) ≤0.0200%
Loss on ignition 50%
Surface area 0.1-0.5 m2/g
Particle size distribution (d50) 15±3µ


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR