n-Tetracosane ≥99%

Supplier: BeanTown Chemical

Synonyms: N-tetracosane

142955-100G 142955-25G 142955-5G
BT142955-100GEA 108.57 USD
BT142955-100G BT142955-25G BT142955-5G
n-Tetracosane ≥99%
n-Tetracosane

CAS: 646-31-1; EC No: 211-474-5; MDL No: MFCD00009352 Crystalline/Powder; Linear Formula: CH3(CH2)22CH3; Molecular Formula: C24H50; MW: 338.66 Melting Point: 49-52°; Boiling Point: 391°; Flash point: 110°C (230°F)


  • Crystalline/Powder

Formula: H₃C(CH₂)₂₂CH₃
Boiling Pt: 391 °C
Melting Pt: 49…52 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00009352
CAS Number: 646-31-1
EINECS: 211-474-5

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR