Polydimethylsiloxane, trimethylsiloxy terminated, M.W. ∼ 400

Supplier: BeanTown Chemical

142925-1L 142925-250ML 142925-50ML
BT142925-1LEA 286 USD
BT142925-1L BT142925-250ML BT142925-50ML
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. ∼ 400
Polydimethylsiloxane, trimethylsiloxy terminated

CAS: 9016-00-6; MDL No: MFCD00148360; RTECS: TQ2690000 Liquid; Linear Formula: (CH3)3SiO(Si(CH3)2O)nSi(CH3)3 ; MW ≈400 Melting Point: -40°; Flash point: 70°C (158°F) Density (g/mL): 0.88; Refractive Index: 1.390


  • RTECS: TQ2690000
  • Refractive index: 1.390
  • Liquid

Formula: (CH₃)₃SiO(Si(CH₃)₂O)nSi(CH₃)₃
Melting Pt: –40 °C
Density: 0.88 g/ml
Flash Pt: 70 °C (158 °F)
MDL Number: MFCD00148360
CAS Number: 9016-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR