Zinc sulfide 99,995

Supplier: BeanTown Chemical

Synonyms: Zinc (II) sulphide, Zinc (II) sulfide

142800-10G 142800-250G 142800-50G
BT142800-10GEA 25 USD
BT142800-10G BT142800-250G BT142800-50G
Zinc sulfide 99,995
Zinc sulphide

CAS: 1314-98-3; EC No: 215-251-3; MDL No: MFCD00011301; RTECS: ZH5400000 3-12mm Pieces; Linear Formula: ZnS; MW: 97.43 Melting Point: 1700° Density (g/mL): 4.1


  • RTECS: ZH5400000
  • 3-12mm Pieces

Formula: SZn
MW: 97.46 g/mol
Melting Pt: 1064
Density: 4.1 g/mL
MDL Number: MFCD00011301
CAS Number: 1314-98-3
EINECS: 215-251-3
Merck Index: 12,10294

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR