Sodium sulfate ≥99.995%
Supplier: BeanTown Chemical
Synonyms:
Disodium sulfate, Bisodium sulfate, Disodium sulphate, Sulfuric acid disodium salt, Bisodium sulphate, Sulphuric acid disodium salt
142715-100G
142715-25G
BT142715-100GEA
642
USD
BT142715-100G
BT142715-25G
Sodium sulfate ≥99.995%
Sodium sulphate
CAS: 7757-82-6; EC No: 231-820-9; MDL No: MFCD00003504; RTECS: WE1650000
Powder; Linear Formula: Na2SO4 ; MW: 142.04
Melting Point: 884°
Density (g/mL): 2.68; Refractive Index: 1.484
Hygroscopic
- RTECS: WE1650000
- Refractive index: 1.484
- Hygroscopic
- Powder
Formula:
Na₂SO₄ MW: 142.04 g/mol Boiling Pt: 1429 Melting Pt: 888 Density: 2.68 g/mL |
MDL Number:
MFCD00003504 CAS Number: 7757-82-6 EINECS: 231-820-9 Merck Index: 12,08829 |
|
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...