Ammonium chloride ≥99.5% ACS
Supplier: BeanTown Chemical
142530-1KG
142530-5KG
BT142530-1KGEA
62
USD
BT142530-1KG
BT142530-5KG
Ammonium chloride ≥99.5% ACS
Ammonium chloride
CAS: 12125-02-9; EC No: 235-186-4; MDL No: MFCD00011420; RTECS: BP4550000
Crystalline; Linear Formula: NH4Cl; MW: 53.49
Melting Point: 340° (sublimes)
Hygroscopic
- RTECS: BP4550000
- Hygroscopic
- Crystalline
Formula:
NH₄Cl Melting Pt: 340 °C (subl.) |
MDL Number:
MFCD00011420 CAS Number: 12125-02-9 EINECS: 235-186-4 |
|
Specification Test Results
Assay | 99.5% min. |
pH of 5% solution at 25°C | 4.5-5.5 |
Residue after ignition | ≤0.01% |
Insoluble matter | ≤0.005% |
Phosphate (PO4) | ≤2ppm |
Heavy metals | ≤5ppm |
Iron (Fe) | ≤2ppm |
Sulfate (SO4) | ≤0.002% |
Calcium (Ca) | ≤0.001% |
Magnesium (Mg) | ≤5ppm |
Clarity of solution | Clear and complete |
Appearance | White crystals |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...