1-Benzyl-4-methylpiperazine hydrochloride ≥98%
Supplier: BeanTown Chemical
142520-10G
142520-50G
BT142520-10GEA
54
USD
BT142520-10G
BT142520-50G
1-Benzyl-4-methylpiperazine hydrochloride ≥98%
1-Benzyl-4-methylpiperazine hydrochloride
CAS: 374898-00-7; MDL No: MFCD03093089
Crystalline/Powder; Molecular Formula: C12H19ClN2; MW: 226.75
- Crystalline/Powder
Formula:
C₁₂H₁₉ClN₂ |
CAS Number:
374898-00-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...