2-Aminofluorene ≥98%

Supplier: BeanTown Chemical

Synonyms: 9H-Fluoren-2-amine, 2-Fluorenamine, Fluoren-2-ylamine

142485-1G 142485-25G 142485-5G
BT142485-1GEA 21 USD
BT142485-1G BT142485-25G BT142485-5G
2-Aminofluorene ≥98%
2-Aminofluorene

CAS: 153-78-6; EC No: 205-817-8; MDL No: MFCD00001125; RTECS: LL5075000 Crystalline; Molecular Formula: C13H11N ; MW: 181.23 Melting Point: 124-128°


  • RTECS: LL5075000
  • Crystalline

Formula: C₁₃H₁₁N
MW: 181.24
Boiling Pt: 327
Melting Pt: 124…128 °C
Density: 1.281
MDL Number: MFCD00001125
CAS Number: 153-78-6
EINECS: 205-817-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR