Sodium formate ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: Formic acid sodium salt

142470-2KG 142470-500G
BT142470-2KGEA 143.33 USD
BT142470-2KG BT142470-500G
Sodium formate ≥99.0% ACS
Sodium formate

  • RTECS: LR0350000
  • Moisture Sensitive
  • Crystalline/Powder

Formula: HCO₂Na
MW: 68.01 g/mol
Melting Pt: 259…262 °C
Density: 1.92 g/mL
MDL Number: MFCD00013101
CAS Number: 141-53-7
EINECS: 205-488-0

Order Now

Specification Test Results

Assay 99.0% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR