Ammonium acetate ≥97%
Supplier: BeanTown Chemical
142455-1KG
142455-250G
142455-5KG
BT142455-1KGEA
73.19
USD
BT142455-1KG
BT142455-250G
BT142455-5KG
Ammonium acetate ≥97%
Ammonium acetate
CAS: 631-61-8; EC No: 211-162-9; MDL No: MFCD00013066; RTECS: AF3675000
Crystalline; Linear Formula: CH3CO2NH4; MW: 77.08
Melting Point: 110-112° (decomposes)
Hygroscopic
- RTECS: AF3675000
- Hygroscopic
- Crystalline
Formula:
H₃CCOONH₄ Melting Pt: 110…112 °C (dec.) |
MDL Number:
MFCD00013066 CAS Number: 631-61-8 EINECS: 211-162-9 |
|
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...