Calcein disodium salt
Supplier: BeanTown Chemical
Synonyms:
Calcein sodium salt
142345-25G
142345-5G
BT142345-25GEA
194
USD
BT142345-25G
BT142345-5G
Calcein disodium salt
Calcein disodium salt
CAS: 108750-13-6; MDL No: MFCD00005049
Powder; Molecular Formula: C30H24N2Na2O13; MW: 666.50
- Powder
Formula:
C₃₀H₂₄N₂Na₂O₁₃ |
MDL Number:
MFCD00082336 CAS Number: 108750-13-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...