Lithium sulfate, anhydrous ≥99.5% (trace metals basis)
Supplier: BeanTown Chemical
142240-1KG
142240-250G
142240-5KG
BT142240-1KGEA
502.65
USD
BT142240-1KG
BT142240-250G
BT142240-5KG
Lithium sulfate, anhydrous ≥99.5% (trace metals basis)
Lithium sulphate
CAS: 10377-48-7; EC No: 233-820-4; MDL No: MFCD00011086
-40 Mesh Powder; Linear Formula: Li2SO4; MW: 109.94
Melting Point: 845°
Density (g/mL): 2.22
Hygroscopic
- Hygroscopic
- -40 Mesh Powder
Formula:
Li₂O₄S MW: 109.95 g/mol Boiling Pt: 1377 Melting Pt: 859 Density: 2.22 g/mL |
MDL Number:
MFCD00011086 CAS Number: 10377-48-7 EINECS: 233-820-4 Merck Index: 12,05569 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...