Calcium fluoride ≥99.95%
Supplier: BeanTown Chemical
142135-1G
142135-25G
142135-5G
BT142135-1GEA
24
USD
BT142135-1G
BT142135-25G
BT142135-5G
Calcium fluoride ≥99.95%
Calcium fluoride
CAS: 7789-75-5; EC No: 232-188-7; MDL No: MFCD00010907; RTECS: EW1760000
Powder; Molecular Formula: CaF2; MW: 78.08
Boiling Point: 2500°
Density (g/mL): 3.18
Hygroscopic
- RTECS: EW1760000
- Hygroscopic
- Powder
Formula:
CaF₂ MW: 78.07 g/mol Boiling Pt: 2500 Melting Pt: 1418 Density: 3.18 g/mL |
MDL Number:
MFCD00010907 CAS Number: 7789-75-5 EINECS: 232-188-7 Merck Index: 13,01669 |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...