Diethylene glycol ≥99%
Supplier: BeanTown Chemical
CAS: 111-46-6; EC No: 203-872-2; MDL No: MFCD00002882; RTECS: ID5950000
Liquid; Linear Formula: (HOCH2CH2)2O; Molecular Formula: C4H10O3; MW: 106.12
Melting Point: -10°; Boiling Point: 245°; Flash point: 143°C (289°F)
Density (g/mL): 1.118; Refractive Index: 1.447
Hygroscopic
- RTECS: ID5950000
- Refractive index: 1.447
- Hygroscopic
- Liquid
Formula:
C₄H₁₀O₃ Boiling Pt: 245 °C Melting Pt: –10 °C Density: 1.118 g/mL Flash Pt: 143 °C (289 °F) |
MDL Number:
MFCD00002882 CAS Number: 111-46-6 EINECS: 203-872-2 |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...