Glycerine 99,5 ACS

Supplier: BeanTown Chemical

Synonyms: Glyceritol, Glysanin, 1,2,3-Propanetriol, Osmoglyn, Glycerin, Glycyl alcohol, Glyrol, Propane-1,2,3-triol, Propanetriol, Trihydroxypropane, Glycerol

142040-1L 142040-4L 142040-500ML
BT142040-1LEA 131.2 USD
BT142040-1L BT142040-4L BT142040-500ML
Glycerine 99,5 ACS
Glycerine

CAS: 56-81-5; EC No: 200-289-5; MDL No: MFCD00004722; RTECS: MA8050000 Liquid; Linear Formula: HOCH2CH(OH)CH2OH ; Molecular Formula: C3H8O3; MW: 92.09 Melting Point: 20°; Boiling Point: 182°/20 mmHg; Flash point: 160°C (320°F) Density (g/mL): 1.25; Refractive Index: 1.474 Hygroscopic


  • RTECS: MA8050000
  • Refractive index: 1.474
  • Hygroscopic
  • Liquid

Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290
Melting Pt: 18.6
Density: 1.25 g/mL
Flash Pt: 160 °C (320 °F)
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5% min.
Neutrality Passes test
Residual water ≤0.5%
Acrolein and glucose Passes test
Chlorinated compounds (as Cl) ≤0.003%
Fatty acid esters (as butyric acid) ≤0.5%
Sulfate (SO4) ≤0.001%
Trace metals (as Pb) ≤2ppm
Residue on evaporation 0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR