5-Amino-2-methoxypyridine ≥98%

Supplier: BeanTown Chemical

141930-25G 141930-5G
BT141930-25GEA 159 USD
BT141930-25G BT141930-5G
5-Amino-2-methoxypyridine ≥98%
5-Amino-2-methoxypyridine

CAS: 6628-77-9; EC No: 229-612-8; MDL No: MFCD00006264; RTECS: US1836000 Liquid; Molecular Formula: C6H8N2O ; MW: 124.14 Melting Point: 29-31°; Boiling Point: 85-90°/1 mmHg; Flash point: 110°C (230°F) Refractive Index: 1.575


  • RTECS: US1836000
  • Refractive index: 1.575
  • Liquid

Formula: C₆H₈N₂O
MW: 124.14
Boiling Pt: 265
Melting Pt: 29…31 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00006264
CAS Number: 6628-77-9
EINECS: 229-612-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR