Tin ≥99.995% (trace metals basis), wire Ø 2.0 mm

Supplier: BeanTown Chemical

141895-100CM 141895-25CM 141895-500CM
BT141895-100CMEA 111 USD
BT141895-100CM BT141895-25CM BT141895-500CM
Tin ≥99.995% (trace metals basis), wire Ø 2.0 mm
Tin

CAS: 7440-31-5; EC No: 231-141-8; MDL No: MFCD00133862 Solid; Molecular Formula: Sn; MW: 118.71 Melting Point: 231.9°; Boiling Point: 2270° Density (g/mL): 7.31


  • Solid

Formula: Sn
Boiling Pt: 2270 °C
Melting Pt: 231.9 °C
Density: 7.31 g/mL
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR