Benzyl alcohol ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol

141880-1L 141880-250ML 141880-4L
BT141880-1LEA 125.36 USD
BT141880-1L BT141880-250ML BT141880-4L
Benzyl alcohol ≥99.0% ACS
Benzyl alcohol

CAS: 100-51-6; EC No: 202-859-9; MDL No: MFCD00004599; RTECS: DN3150000 Liquid; Molecular Formula: C7H8O ; MW: 108.14 Melting Point: -16°; Boiling Point: 203-205°; Flash point: 96°C (205°F) Density (g/mL): 1.045; Refractive Index: 1.539


  • RTECS: DN3150000
  • Refractive index: 1.539
  • Liquid

Formula: C₆H₅CH₂OH
MW: 108.14 g/mol
Boiling Pt: 203…205 °C
Melting Pt: –16 °C
Density: 1.045 g/mL
Flash Pt: 96 °C (205 °F)
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0% min.
Acetophenone ≤0.02%
Benzaldehyde ≤0.01%
Color (APHA) ≤20
Residue after ignition ≤0.005%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR