n-Heptadecane ≥99%

Supplier: BeanTown Chemical

141715-100G 141715-25G 141715-500G
BT141715-100GEA 57 USD
BT141715-100G BT141715-25G BT141715-500G
n-Heptadecane ≥99%
n-Heptadecane

CAS: 629-78-7; EC No: 211-108-4; MDL No: MFCD00009002 Crystalline; Linear Formula: CH3(CH2)15CH3; Molecular Formula: C17H36; MW: 240.48 Melting Point: 20-22°; Boiling Point: 302°; Flash point: 149°C (300°F) Density (g/mL): 0.777; Refractive Index: 1.436


  • Refractive index: 1.436
  • Crystalline

Formula: H₃C(CH₂)₁₅CH₃
Boiling Pt: 302 °C
Melting Pt: 20…22 °C
Density: 0.777 g/mL
Flash Pt: 149 °C (300 °F)
MDL Number: MFCD00009002
CAS Number: 629-78-7
EINECS: 211-108-4

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR