L(-)-Glutathione (reduced form) ≥97%
Supplier: BeanTown Chemical
Synonyms:
L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced
141690-100G
141690-25G
141690-5G
BT141690-100GEA
646.17
USD
BT141690-100G
BT141690-25G
BT141690-5G
L(-)-Glutathione (reduced form) ≥97%
L(-)-Glutathione (reduced form)
CAS: 70-18-8; EC No: 200-725-4; MDL No: MFCD00065939; RTECS: MC0556000
Powder; Molecular Formula: C10H17N3O6S; MW: 307.33
Melting Point: 192-195° (decomposes)
- RTECS: MC0556000
- Powder
Formula:
C₁₀H₁₇N₃O₆S Melting Pt: 192…195 °C (dec.) |
MDL Number:
MFCD00065939 CAS Number: 70-18-8 EINECS: 200-725-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...