1-Octadecene ≥90%, Technical Grade
Supplier: BeanTown Chemical
141680-1L
141680-250ML
BT141680-1LEA
46.6
USD
BT141680-1L
BT141680-250ML
1-Octadecene ≥90%, Technical Grade
1-Octadecene
CAS: 112-88-9; EC No: 204-012-9; MDL No: MFCD00009003
Liquid; Linear Formula: CH3(CH2)15CH=CH2; Molecular Formula: C18H36O3; MW: 252.48
Melting Point: 14-16°; Boiling Point: 179°/15 mmHg; Flash point: 149°C (300°F)
Density (g/mL): 0.789; Refractive Index: 1.444
- Refractive index: 1.444
- Liquid
Formula:
C₁₈H₃₆ MW: 252.48 g/mol Boiling Pt: 315 Melting Pt: 17.5 Density: 0.789 g/mL Flash Pt: 149 °C (300 °F) |
MDL Number:
MFCD00009003 CAS Number: 112-88-9 EINECS: 204-012-9 |
Specification Test Results
Assay | 90% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...