3-Nitrophthalic acid ≥95%

Supplier: BeanTown Chemical

141615-100G 141615-500G
BT141615-100GEA 55 USD
BT141615-100G BT141615-500G
3-Nitrophthalic acid ≥95%
3-Nitrophthalic acid

CAS: 603-11-2; EC No: 210-030-8; MDL No: MFCD00007138 Powder; Molecular Formula: C8H5NO6 ; MW: 211.13 Melting Point: 210° (decomposes)


  • Powder

Formula: C₈H₅NO₆
MW: 211.13 g/mol
Boiling Pt: 441
Melting Pt: 210 °C (dec.)
MDL Number: MFCD00007138
CAS Number: 603-11-2
EINECS: 210-030-8

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR