7-Azaindole-5-boronic acid pinacol ester ≥97%
Supplier: BeanTown Chemical
Synonyms:
7-Azaindole-5-boronic Acid Pinacol Ester
141555-1G
141555-250MG
BT141555-1GEA
311
USD
BT141555-1G
BT141555-250MG
7-Azaindole-5-boronic acid pinacol ester ≥97%
7-Azaindole-5-boronic acid pinacol ester
CAS: 754214-56-7; MDL No: MFCD08060937
Solid; Molecular Formula: C13H17BN2O2 ; MW: 244.10
Melting Point: 243-248°
Air Sensitive
- Air Sensitive
- Solid
Formula:
C₁₃H₁₇BN₂O₂ MW: 244.101 g/mol Melting Pt: 243…248 °C |
MDL Number:
MFCD08060937 CAS Number: 754214-56-7 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...