Cobalt ≥99.995% (trace metals basis), rods Ø 5 mm
Supplier: BeanTown Chemical
141450-150MM
141450-50MM
BT141450-150MMEA
1291
USD
BT141450-150MM
BT141450-50MM
Cobalt ≥99.995% (trace metals basis), rods Ø 5 mm
Cobalt
CAS: 7440-48-4; EC No: 231-158-0; MDL No: MFCD00010935
Solid; Molecular Formula: Co; MW: 58.93
Melting Point: 1495°; Boiling Point: 2900°
Density (g/mL): 8.92
- Solid
Formula:
Co MW: 58.93 g/mol Boiling Pt: 2870 Melting Pt: 1495 Density: 8.92 g/mL |
MDL Number:
MFCD00010935 CAS Number: 7440-48-4 EINECS: 231-158-0 Merck Index: 12,02488 |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...