(±)-1,2-Propanediol ≥98%
Supplier: BeanTown Chemical
CAS: 57-55-6; EC No: 200-338-0; MDL No: MFCD00064272; RTECS: TY2000000
Liquid; Linear Formula: CH3CH(OH)CH2OH; Molecular Formula: C3H8O2; MW: 76.09
Melting Point: -60°; Boiling Point: 187°; Flash point: 103°C (217°F)
Density (g/mL): 1.036; Refractive Index: 1.432
Hygroscopic
- RTECS: TY2000000
- Refractive index: 1.432
- Hygroscopic
- Liquid
Formula:
CH₃CH(OH)CH₂OH Boiling Pt: 187 °C Melting Pt: –60 °C Density: 1.036 g/mL Flash Pt: 103 °C (217 °F) |
MDL Number:
MFCD00064272 CAS Number: 57-55-6 EINECS: 200-338-0 |
|
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...