Hafnium ≥99.5% (trace metals basis excluding Zr), foil, annealed, Thickness 1.0 mm
Supplier: BeanTown Chemical
140675-100X100
140675-50X50MM
BT140675-100X1EA
1264
USD
BT140675-100X1
BT140675-50X50
Hafnium ≥99.5% (trace metals basis excluding Zr), foil, annealed, Thickness 1.0 mm
Hafnium
CAS: 7440-58-6; EC No: 231-166-4; MDL No: MFCD00011032
Solid; Molecular Formula: Hf; MW: 178.49
Melting Point: 2227°; Boiling Point: 4602°
Density (g/mL): 13.3
- Solid
Formula:
Hf Boiling Pt: 4602 °C Melting Pt: 2227 °C Density: 13.3 g/mL |
MDL Number:
MFCD00011032 CAS Number: 7440-58-6 EINECS: 231-166-4 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...