Tellurium ≥99.9999% (trace metals basis), shot, Diameter 2-5 mm
Supplier: BeanTown Chemical
140495-100G
140495-25G
140495-5G
BT140495-100GEA
474
USD
BT140495-100G
BT140495-25G
BT140495-5G
Tellurium ≥99.9999% (trace metals basis), shot, Diameter 2-5 mm
Tellurium
CAS: 13494-80-9; EC No: 236-813-4; MDL No: MFCD00134062; RTECS: WY2625000
Solid; Molecular Formula: Te; MW: 127.60
Melting Point: 450°; Boiling Point: 990°
Density (g/mL): 6.24
- RTECS: WY2625000
- Solid
Formula:
Te Boiling Pt: 990 °C Melting Pt: 450 °C Density: 6.24 g/mL |
MDL Number:
MFCD00134062 CAS Number: 13494-80-9 EINECS: 236-813-4 UN: 3284 ADR: 6.1,III |
Specification Test Results
Assay | 99.9999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...