Stearic acid ≥90%, Technical Grade
Supplier: BeanTown Chemical
Synonyms:
Octadecanoic acid, Octadecylic acid
140210-1KG
140210-5KG
BT140210-1KGEA
32.44
USD
BT140210-1KG
BT140210-5KG
Stearic acid ≥90%, Technical Grade
Stearic acid
CAS: 57-11-4; EC No: 200-313-4; MDL No: MFCD00002752; RTECS: WI2800000
Flakes; Linear Formula: CH3(CH2)16COOH; Molecular Formula: C18H36O2; MW: 284.48
Melting Point: 67-72°; Boiling Point: 361°; Flash point:
- RTECS: WI2800000
- Flakes
Formula:
C₁₈H₃₆O₂ Boiling Pt: 361 °C Melting Pt: 67…72 °C Density: 0.845 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00002752 CAS Number: 57-11-4 EINECS: 200-313-4 |
|
Specification Test Results
Assay | 90% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...