Sodium chloride ≥99.999% (trace metals basis)

Supplier: BeanTown Chemical

140175-100G 140175-25G 140175-500G
BT140175-100GEA 239.22 USD
BT140175-100G BT140175-25G BT140175-500G
Sodium chloride ≥99.999% (trace metals basis)
Sodium chloride

CAS: 7647-14-5; EC No: 231-598-3; MDL No: MFCD00003477; RTECS: VZ4725000 Solid; Linear Formula: NaCl; MW: 58.44 Melting Point: 801°; Boiling Point: 1413° Density (g/mL): 2.165; Refractive Index: 1.544


  • RTECS: VZ4725000
  • Refractive index: 1.544
  • Solid

Formula: NaCl
Boiling Pt: 1413 °C
Melting Pt: 801 °C
Density: 2.165 g/mL
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR