4-(Trifluoromethyl)phenylboronic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-(Trifluoromethyl)benzeneboronic acid, (4-(Trifluoromethyl)phenyl)metaboric acid, Dihydroxy(4-(trifluoromethyl)phenyl)borane, (4-(Trifluoromethyl)phenyl)boronic acid

140170-1G 140170-25G 140170-5G
BT140170-1GEA 69.32 USD
BT140170-1G BT140170-25G BT140170-5G
4-(Trifluoromethyl)phenylboronic acid ≥98%
4-(Trifluoromethyl)phenylboronic acid

CAS: 128796-39-4; MDL No: MFCD00151855 Powder; Molecular Formula: C7H6BF3O2 ; MW: 189.93 Melting Point: 245-250°


  • Powder

Formula: C₇H₆BF₃O₂
MW: 189.93 g/mol
Melting Pt: 245…250 °C
MDL Number: MFCD00151855
CAS Number: 128796-39-4

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR