Silica gel, Pore size: 60 Å 0.019-0.037mm (400-600 mesh)

Supplier: BeanTown Chemical

140010-1KG 140010-250G 140010-50G
BT140010-1KGEA 272.51 USD
BT140010-1KG BT140010-250G BT140010-50G
Silica gel, Pore size: 60 Å 0.019-0.037mm (400-600 mesh)
Silica gel

CAS: 63231-67-4; EC No: 231-545-4; MDL No: MFCD00011232 Powder, S.A. 500-600 m2/g Melting Point: 1600°; Boiling Point: 2230° Hygroscopic


  • Hygroscopic
  • Powder, S.A. 500-600 m²/g

Formula: SiO₂
Boiling Pt: 2230 °C
Melting Pt: >1600 °C
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR