Chromium(III) oxide ≥99.5%
Supplier: BeanTown Chemical
Synonyms:
Chromic oxide
139720-100G
139720-25G
BT139720-100GEA
179.55
USD
BT139720-100G
BT139720-25G
Chromium(III) oxide ≥99.5%
Chromium (III) oxide
CAS: 1308-38-9; EC No: 215-160-9; MDL No: MFCD00010949; RTECS: GB6475000
3-6mm Fused Lump; Molecular Formula: Cr2O3; MW: 151.99
- RTECS: GB6475000
- Fused lump
Formula:
Cr₂O₃ MW: 151.99 g/mol Boiling Pt: 4000 Melting Pt: 2435 |
MDL Number:
MFCD00010949 CAS Number: 1308-38-9 EINECS: 215-160-9 Merck Index: 13,02247 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...